missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Captair 531 Midcap Filtering Storage Cabinet

The Captair 531 Midcap filtering chemical storage cabinet provides outstanding filtration quality to ensure that users and laboratory technicians are protected against chemical hazards. Users are given the best possible view of the stored products through their transparent doors and beneficiate in proximity of its daily chemical products, thanks to its small size.

Brand:  Erlab™ AB62

Additional Details : Weight : 15.00000kg

Product Code. 17901692

  • £650.81 / Each
Estimated Shipment: 19-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Air Flow: 10 m3/hr.
  • Structure made of corrosion resistant electro-galvanized steel coated with anti-acid polymer
  • Door is clear and made with chemical resistant acrylic for easy viewing
  • Filtration module is made with injected polypropylene
  • Carbon filtration for gases and vapors: AS for organic vapors, BE for acid vapors, F for formaldehyde vapors, K for ammonia vapors
  • 1 storage compartment
  • Delivered with 1 shelf and 1 tray
  • 2 absorbent mats
  • Storage capacities: About 72 x 20 mL bottles or 10 x 500 mL, depending of the type of container used
Specifications

Specifications

Chemicals Cabinet
9.5 in.
242 mm
19.9 in.
506 mm
1
CE, AFNOR NF X 15 211:2009
16 in.
Steel, Polymer
506 x 242 x 406 mm
White
406 mm
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers