missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Captair 632A Smart Filtering Storage Cabinet

The Captair 632A Midcap filtering chemical storage cabinet provides outstanding filtration quality to ensure that users and laboratory technicians are protected against chemical hazards. Users are given the best possible view of the stored products through their transparent doors and beneficiate in proximity of its daily chemical products, thanks to its small size.

Brand:  Erlab™ AB72

Additional Details : Weight : 30.00000kg

Product Code. 17981692

  • £867.74 / Each
Estimated Shipment: 19-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Air Flow: 10 m3/hr.
  • Structure made of corrosion resistant electro-galvanized steel coated with anti-acid polymer
  • Door is clear and made with chemical resistant acrylic for easy viewing
  • Filtration module is made with injected polypropylene
  • Carbon filtration for gases and vapors: AS for organic vapors, BE for acid vapors, F for formaldehyde vapors, K for ammonia vapors
  • 1 storage compartment
  • Delivered with 1 shelf and 1 tray
  • 2 absorbent mats
  • Storage capacities: About 24 x 500 mL bottles (high shelves) or 20 x 1 L bottles (low shelves), depending on the type of container used
  • Key lock
Specifications

Specifications

Chemicals Cabinet
18.6 in.
471.2 mm
23.7 in.
603 mm
1
CE, AFNOR NF X 15 211:2009
27 in.
Steel, Polymer
603 x 471.2 x 685 mm
White
685 mm
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers