missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Captair 632B Midcap Filtering Storage Cabinets

These Captair 632 B Midcap filtering chemical storage cabinets provide outstanding filtration quality that ensure users and laboratory technicians are protected against chemical hazards. The technology adapts to the type of product being stored and the size is ideal for an under-bench localization. Users are given the best possible view of the stored products through their transparent doors.

Brand:  Erlab™ AB82//04

Additional Details : Weight : 42.00000kg

 View more versions of this product

Product Code. 17991692

  • £1338.92 / Each
Estimated Shipment: 14-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Air Flow: 11 m3/hr.
  • Structure made of corrosion resistant electro-galvanized steel coated with anti-acid polymer
  • Doors are clear and made with chemical resistant acrylic for easy viewing
  • Filtration module is made with injected polypropylene
  • Carbon filtration: AS for organic vapors, BE for acid vapors, F for formaldehyde vapors, K for ammonia vapors and particulate filtration for powders (option)
  • HEPA H13: 99.95% efficiency filtration of particles over 0.1 μm in size
  • Key lock
Specifications

Specifications

Chemicals Cabinet
23 in.
1 Tray, 2 Absorbent Mats, 1 Storage Compartment, Storage for 25 x 1 L or 35 x 100 mL or 70 x 50 mL Bottles
25.7 in.
Sliding
1
CE, AFNOR NF X 15-211:2009, EN 1822:1998
28.1 in.
Steel, Polymer
654 x 583.6 x 715 mm
White
Hinged
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers