missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Captair 832BF Midcap Filtering Storage Cabinet

The filtering chemical storage cabinet Captair 832 BF Midcap provides outstanding filtration quality by ensuring users and laboratory technicians are protected against chemical hazards. Their technology adapts to the type of product being stored. Users are given the best possible view of the stored products through their transparent doors.

Brand:  Erlab™ AB32

Additional Details : Weight : 110.00000kg

Product Code. 17991672

  • £2145.92 / Each
Estimated Shipment: 03-09-2024
to see stock.

Description

Description

  • Air Flow: 70 m3/hr.
  • Structure made of corrosion resistant electro-galvanized steel coated with anti-acid polymer
  • Doors made of clear, chemical resistant acrylic for easy viewing
  • Filtration module is made with injected polypropylene
  • Carbon filtration: AS for organic vapors, BE for acid vapors, F for formaldehyde vapors, K for ammonia vapors
  • Particulate filtration for powders: HEPA H14:99.995% efficiency filtration of particles over 0.1 μm in size
  • Double doors with shelves
  • Delivered with 10 movable shelves with integrated retention tray
  • 2 absorbing mats
  • 2 storage compartments
  • 120 x 1 L glass bottles for storage
  • Key lock
  • List of approved chemicals
Specifications

Specifications

Chemicals Cabinet
35.4 in.
900 mm
19.7 in.
500 mm
10
CE, AFNOR NF X 15-211:2009, EN 1822:1998
81.1 in.
Steel, Polymer
500 x 900 x 2060 mm
White
2060 mm
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers