missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erlab™ Captair 834 Smart Filtering Storage Cabinets
These filtering chemical storage cabinets provide outstanding filtration quality by ensuring that users and laboratory technicians are protected against chemical hazards. Their technology adapts to the type of product being stored. Users are given the best possible view of the stored products through their shelves, transparent doors and are alerted in the event of prolonged opening.
£2994.57 - £4227.63
Specifications
Certifications/Compliance | CE, AFNOR NF X 15 211:2009, EN 1822:1998 |
---|---|
Color | White |
Depth (English) Exterior | 26 in. |
Dimensions (D x W x H) | 660 x 900 x 2140 mm |
Height (English) Exterior | 84.3 in. |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17901702
|
Erlab™
AB41//03 |
Integrated Retention Tray, Storage for 100 x 1 L Glass Bottles |
£2994.57
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17971662
|
Erlab™
AB41//04 |
8 Fixed Trays, Storage for 120 x 1 L Glass Bottles |
£4227.63
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Air Flow: 220 m3/hr. (129 CFM)
- Structure made of corrosion resistant electro-galvanized steel coated with anti-acid polymer
- Doors made of clear, chemical resistant acrylic for easy viewing
- Filtration module is made with injected polypropylene
- Filtration technology: 1 column can be configured to handle liquids, powders, or both
- Following filtration column configuration: Carbon filtration for gases and vapors and particulate filtration for powders
- Detection sensor for Type S, for solvents; Type A, for acids; Type F, for formaldehydes
- Door sensor alarm deploys if doors are left open
- 2 storage compartments
- Absorbing mats: 2
- Key lock
- List of approved chemicals
- Communication Interface: When the light is pulsing: doors left open, containment is compromised, filter breakthrough
- eGuard APP: For remote control to monitor the status of the storage cabinet, change the settings, and deliver safety alerts immediately to your devices
Specifications
CE, AFNOR NF X 15 211:2009, EN 1822:1998 | |
26 in. | |
84.3 in. | |
Chemicals Cabinet | |
2 |
White | |
660 x 900 x 2140 mm | |
Steel, Polymer | |
35.4 in. |