Learn More
Erlab™ Captair by Erlab™ Stainless Steel Work Surfaces
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Stainless Steel Work Surfaces are for use with Captair PCR work stations.
Brand: Erlab™ I0118050001
Code : NEW
Additional Details : Weight : 9.00000kg
Description
- High chemical and mechanical resistance
- Rounded corners to facilitate cleaning operations
- Built in spill tray
Specifications
Stainless Steel | |
CaptairFlow 483 ductless fume hoods |