Learn More
Erlab™ Captair™ Mobile Ductless Fume Hood Filters
Provides personal protection while handling liquid, gaseous and solid chemicals. Erlab™ Captair™ Mobile Ductless Fume Hood Filters provide filtration of organic, acid, formadehyde, and ammonia vapors as well as fine powders.
Brand:  Erlab™ H11074101
Code : 50
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12010393
  • £498.00 / Each
Estimated Shipment: 18-07-2024
to see stock.
Description
Description
Provides personal protection during handlings of liquid, gaseous and solid chemicals.
Filter types available:
  • GF4 AS: Oganic vapors
  • GF4 BE +: Organix vapors plus acid vapors
  • GF4 F: Formaldehyde vapors
  • GF4 K: Ammonia vapors
  • GF4 HP: HI4 HEPA filter for powders
Specifications
Specifications
AS
Mobile Ductless Filtration Fume Hood
Replacement Filter
For Organic Vapors