Learn More
Erlab™ Captair Smart Trespa™ Work Surface
Erlab™ Captair Smart Trespa™ Work Surface is designed to be used with the range of Captair Smart Ductless Filtering Fume Hoods.
Brand: Erlab™ I0162030001
This item is not returnable.
View return policy
Description
- Made of phenolic resin with ergonomic arm for a comfortable working position
- High chemical and mechanical resistance
- Ideal for precision weighing operations
Specifications
Smart 481 Fume Hood | |
545 mm | |
Work Surface |
Spill Tray | |
1190 mm | |
Phenolic Resin |
Product Content Correction
Product Title
Erlab™ Captair Smart Trespa™ Work Surface > For Use With: Smart 481 Fume Hood
Spot an opportunity for improvement?Share a Content Correction