Learn More
Erlab™ Captair Smart Trespa™ Work Surface
Erlab™ Captair Smart Trespa™ Work Surface is designed to be used with the range of Captair Smart Ductless Filtering Fume Hoods.
Brand:  Erlab™ I0163030001
 View more versions of this product
Product Code. 15515164
  • £1315.00 / Each
Explore more special offers
Description
Description
  • Made of phenolic resin with ergonomic arm for a comfortable working position
  • High chemical and mechanical resistance
  • Ideal for precision weighing operations
Specifications
Specifications
Smart 392 Fume Hood
644 mm
Work Surface
Spill Tray
875 mm
Phenolic Resin
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Erlab™ Captair Smart Trespa™ Work Surface > For Use With: Smart 392 Fume Hood