missing translation for 'onlineSavingsMsg'
Learn More

Erlab Storage cabinet 834 800mm x 605mm x 2175/2255mm vented, Flex tech, steel, 100% polyester coated

Brand:  Erlab B01PAO

Storage cabinet 834 800mm x 605mm x 2175/2255mm vented, Flex tech, steel, 100% polyester coated

Code : 50

Additional Details : Weight : 1.00000kg

Product Code. 12898713

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers