missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ HiRes Ion Exchange Chromatography Columns

Prepacked columns for high-resolution purification of proteins and other biomolecules by ion exchange chromatography. They are well-suited for the preparation of highly pure proteins for structural studies and protein analysis.

Brand:  Cytiva 29275879

Additional Details : CAS Number : 64-17-5 Weight : 0.20000kg


 View more versions of this product

Product Code. 16277791

  • £5267.00 / Each
Estimated Shipment: 27-08-2024
to see stock.
Add to basket

Description

Description

  • High resolution columns for laboratory scale ion exchange chromatography
  • Suitable for preparing highly pure proteins, and protein analysis in research or quality control applications
  • High quality column with excellent reproducibility, durability, and high chemical and pH stabilities
  • Replacement for Mono Q™ and Mono S™ columns delivering similar or improved resolution, with easy transition from one column platform to the other
  • Chromatography packing resins are based on a high-flow agarose base matrix and a small bead size (9 μm average)
Specifications

Specifications

100 mm
Ionic Capacity: ∼0.12 mmol H+/mL resin
0.5 to 2 mL/min.
Methyl Sulfonate
∼9μm
Capto™ HiRes S 10/100 Column
Highly Cross-Linked Agarose
2 to 12
4°C to 40°C
Ion Exchange Column
8 mL
Anaylsis or Purification of Proteins and Nucleic Acids
4 MPa (40 bar)
Capto
Tricorn™ Glass
Spherical
1 Ea.
10 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Capto HiRes S 10/100

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers