missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Capto™ Multimodal (MMC) Chromatography Media
Used to solve specific purification challenges at both high and low ionic strengths
£174.00 - £425.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10681521
|
Cytiva
17-5317-10 |
25 mL |
£174.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10089269
|
Cytiva
17-5317-02 |
100 mL |
£425.00
100mL |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
Description
- Composed of rigid, high-flow agarose matrix and novel multimodal ligand
- Used to bind proteins at conductivity of feed material
- Ligand contains carboxylic acid group and exhibit the properties of weak cation exchanger
- High dynamic binding capacity at high conductivity
- High-volume throughput cuts process times
- Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
- Dynamic binding capacity at 10% breakthrough as measured at a residence time of 2 min, 300cm/h in a Tricorn(TM) 5/100 column with 10cm bed height in 50mM Sodium acetate, pH 4.75, 250mM NaCl.
Warranty
Store at 4° to 30°C in 20% Ethanol
Specifications
Capto MMC Media | |
75 μm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Capto MMC, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.