missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Capto Q ImpRes, 25mL
Designed to meet the demands of modern, large-scale manufacturers. Cytiva Capto Q ImpRes, 25ml is a strong anion exchange BioProcess medium that enables fast, efficient, and cost-effective intermediate and polishing protein purification.
Brand: Cytiva 17-5470-10
This item is not returnable.
View return policy
Description
Capto Q ImpRes is a BioProcess chromatography medium developed for high-throughput intermediate purification and polishing of a wide range of biopharmaceutical proteins. It is part of an expanded high-res platform based on the Capto product line with traditional ionic group ligands. By combining the high-flow characteristics of agarose Capto media with a small particle size, Capto Q ImpRes delivers excellent pressure-flow properties with impressive resolution and improved productivity. The ability to run at higher flow rates and higher bed heights also increases flexibility in process design.- High-resolution purification and polishing—based on the well-established Capto platform
- Flexible process design—large operational window of flow rates and bed heights
- High-throughput purifications—easy to optimize and scale up
- Higher manufacturing productivity enables improved process economy
- Security of supply and comprehensive technical and regulatory support
Specifications
Capto Q ImpRes, 25 mL | |
Bioprocess Chromatography | |
4°C to 30°C in 20% Ethanol | |
25 mL | |
2 to 12 |
High-flow agarose | |
Quaternary amine | |
>55 mg BSA, >48 mg β-lactoglobulin/mL Chromatography Medium | |
36 to 44 μm (d50V) | |
25 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Capto Q ImpRes, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.