missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Capto S™ ImpAct Chromatography Media
Capto S ImpAct is a strong cation exchange chromatography medium (resin) developed for polishing of monoclonal antibodies (MAbs) and other biomolecules.
£175.00 - £515.00
Specifications
Product Type | Capto S ImpAct Chromatography Media |
---|---|
Material | High-flow agarose |
pH Range | 4 to 12 |
Particle Size | 50μm |
Matrix | Ion Exchange Chromatography Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15341642
|
Cytiva
17371701 |
25mL |
£175.00
25mL |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
15351642
|
Cytiva
17371702 |
100mL |
£515.00
100mL |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
Description
- High binding capacity, > 100 mg MAb/mL medium.
- Efficient aggregate removal at high load of monoclonal antibodies.
- High-resolution polishing based on the well-established Capto platform.
- Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling.
- High productivity enabling cost-efficient manufacturing.
- Security of supply and comprehensive regulatory support.
Specifications
Capto S ImpAct Chromatography Media | |
4 to 12 | |
Ion Exchange Chromatography Media |
High-flow agarose | |
50μm |