missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Capto™ SP ImpRes BioProcess Media
A strong cation exchange BioProcess medium for efficient and cost-effective protein purification.
Brand: Cytiva 17546810
This item is not returnable.
View return policy
Description
- Chemically stable with commonly used aqueous buffers
- High manufacturing productivity
- Sulfate group ligand
- High-flow agarose matrix
- Flexible process design due to large operational window of flow rates and bed heights
Avoid using oxidizing agents and cationic detergents
Developed and supported for large-scale chromatography; used for high-throughput intermediate purification and polishing steps of a wide range of biomolecules
Certifications
Certificate of Analysis
Specifications
25 mL | |
For Large Scale PurifiCatIon of Proteins | |
>70 mg Lysozyme, >95 mg BSA | |
220 cm/Hoμr | |
7.87 in. | |
0.13 to 0.16 mM Hl+/mL | |
Strong CatIon Exchanger | |
High-flow Agarose | |
20% Ethanol, 0.2 M Sodium Acetate |
1 M NaOH, 8 M Urea, 6 M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol | |
4°C to 30°C | |
Capto SP ImpRes Media | |
36 to 44 μm | |
20 cm | |
Certificate of Analysis | |
300 kPa | |
3 to 14 | |
Sulphonate Group |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Capto SP ImpRes, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol