missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Captopril, 98%

£43.22 - £441.20

Chemical Identifiers

CAS 62571-86-2
Molecular Formula C9H15NO3S
Molecular Weight (g/mol) 217.28
MDL Number MFCD00168073
InChI Key FAKRSMQSSFJEIM-RQJHMYQMSA-N
Synonym captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril
PubChem CID 44093
ChEBI CHEBI:3380
IUPAC Name (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
SMILES CC(CS)C(=O)N1CCCC1C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10295771
View Documents Promotion Details
Thermo Scientific Chemicals
343700010
1 g Glass bottle
£43.22
1g
Please to purchase this item. Need a web account? Register with us today!
10205781
View Documents Promotion Details
Thermo Scientific Chemicals
343700050
5 g Glass bottle
£130.65
5g
Please to purchase this item. Need a web account? Register with us today!
10154061
View Documents Promotion Details
Thermo Scientific Chemicals
343700250
25 g Glass bottle
£441.20
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62571-86-2
217.28
FAKRSMQSSFJEIM-RQJHMYQMSA-N
44093
(2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
C9H15NO3S
MFCD00168073
captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril
CHEBI:3380
CC(CS)C(=O)N1CCCC1C(=O)O

Specifications

62571-86-2
White
97.5% min. (HPLC)
MFCD00168073
− 129.50
Solubility in water: soluble. Other solubilities: freely soluble in alcohol,chloroform,methylenech,loride
CC(CS)C(=O)N1CCCC1C(=O)O
217.28
CHEBI:3380
98%
Captopril, 98%
103.0°C to 108.0°C
Authentic
C9H15NO3S
15, 1776
captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril
FAKRSMQSSFJEIM-RQJHMYQMSA-N
(2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
44093
217.28
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Causes serious eye damage.
May damage the unborn child.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 263-607-1

missing translation for 'rtecsNumber' : UY0550000

Product Identifier
  • Captopril

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only