Learn More
Thermo Scientific™ CaptureSelect™ AAVX Ligand Leakage ELISA
The CaptureSelect AAVX Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAVX Affinity Matrix.
Brand: Thermo Scientific™ 810352210
Additional Details : Weight : 0.03200kg
Description
The CaptureSelect AAVX Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAVX Affinity Matrix. The ligand, a 13-kD recombinant protein, is covalently coupled for high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the purified AAV. This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. POROS CaptureSelect AAVX Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
3 x 1 mL, store at -5°C to -30°C. | |
ELISA Kit | |
Dry Ice |
CaptureSelect™ | |
10 Assays |