Learn More
Carbamates Standard solution 10 components, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical F114161
Code : 04
Additional Details : Weight : 0.00080kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Specifications
Ten Components Carbamates Standard Solution | |
Ampoule | |
Organic Multi-component Standard Solution | |
Aldicarb (CAS:116-06-3), Aldicarb-sulfone (CAS:1646-88-4), Aldicarb-sulfoxide (CAS:1646-87-3), Propoxur (baygon) (CAS:114-26-1), Carbaryl (CAS:63-25-2), Carbofuran (CAS:1563-66-2), Carbofuran-3-hydroxy (CAS:16655-82-6), Methiocarb (CAS:2032-65-7), Methomyl (CAS:16752-77-5), Oxamyl (CAS:23135-22-0) |
100μg/mL | |
1 mL | |
1230 | |
Methanol |
Safety and Handling
- Standard solution 10 Components in methanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H370-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P330-Rinse mouth.
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use to extinguish.
- P501-Dispose of contents/container to .