missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Carbamazepine

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Descripción
Inhibitor of neuronal voltage-gated Na+ channels. Exhibits anticonvulsant activity. Potentiates GABA-induced Cl- currents in HEK 293 cells expressing the GABAA receptor α1β2γ2 subtype combination. Can induce autophagy by inhibiting inositol synthesis. Also delays disease onset and prolongs survival in a mouse amyotrophic lateral sclerosis (ALS) model, as well as reducing motor neuron loss and altered muscle morphology.
Especificaciones
Especificaciones
| CAS | 298-46-4 |
| Quantity | 50 mg |
| Molecular Formula | C15H12N2O |
| Formula Weight | 236.27 |
| Purity | >99% |
| Molecular Weight (g/mol) | 236.27 |
| Chemical Name or Material | Carbamazepine |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido