missing translation for 'onlineSavingsMsg'
Learn More

Carbamylcholine chloride, 98+%

Product Code. p-4228989 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10331753

Brand: Thermo Scientific Acros 108240050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51-83-2 | C6H15ClN2O2 | 182.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-83-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012011
InChI Key AIXAANGOTKPUOY-UHFFFAOYSA-N
Synonym carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
PubChem CID 5831
ChEBI CHEBI:3385
IUPAC Name 2-carbamoyloxyethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CCOC(N)=O

Specifications

Melting Point 200°C to 204°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99%
Linear Formula H2NCO2CH2CH2N(CH3)3Cl
Merck Index 15,1781
Solubility Information Solubility in water: 1g/mL. Other solubilities: 1g/50 mL alcohol,1g/10 mL methanol,practically insoluble in chloroform and ether
Formula Weight 182.65
Percent Purity 99%
Infrared Spectrum Authentic
Loss on Drying 2.0% max. (105°C, 2 hrs)
Packaging Glass bottle
Chemical Name or Material Carbamylcholine chloride
Show More Show Less
compliance-icons
Product Identifier
  • Carbamylcholine chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.