Learn More
Carbenicillin (Disodium Salt), Fisher BioReagents
£44.10 - £88.60
Chemical Identifiers
CAS | 4800-94-6 |
---|---|
Molecular Formula | C17H16N2Na2O6S |
Molecular Weight (g/mol) | 422.36 |
MDL Number | MFCD00077683 |
InChI Key | RTYJTGSCYUUYAL-YCAHSCEMSA-L |
Synonym | α-Carboxybenzylpenicillin |
PubChem CID | 20933 |
ChEBI | CHEBI:34609 |
IUPAC Name | disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
SMILES | [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10438923
|
Fisher BioReagents
BP2648-250 |
250 mg | Amber Glass |
£44.10
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10163193
|
Fisher BioReagents
BP2648-1 |
1 g | Amber Glass |
£88.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10396833
|
Fisher BioReagents
BP2648-5 |
5 g | Amber Glass |
£50.05
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Used for the selection of ampicillin-resistant transformed cells and for the study of penicillin-sensitive transpeptidases in cell wall biosynthesis. Antimicrobial spectrum: Gram-positive and Gram-negative bacteria, Pseudomonas.Chemical Identifiers
4800-94-6 | |
422.36 | |
RTYJTGSCYUUYAL-YCAHSCEMSA-L | |
20933 | |
disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
C17H16N2Na2O6S | |
MFCD00077683 | |
α-Carboxybenzylpenicillin | |
CHEBI:34609 | |
[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O |
Specifications
Carbenicillin | |
4800-94-6 | |
≥92 % | |
MFCD00077683 | |
α-Carboxybenzylpenicillin | |
[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O | |
422.36 | |
CHEBI:34609 | |
92% min. | |
White | |
Carbenicillin, Disodium Salt |
Solid | |
200°C | |
C17H16N2Na2O6S | |
770μg/mg min. | |
RTYJTGSCYUUYAL-YCAHSCEMSA-L | |
disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
20933 | |
422.4 | |
6% Max. | |
738°C |
Safety and Handling
WARNING!
Emergency Overview
May cause allergic respiratory and skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.
NFPA
Health:2
Flammability:0
Instability:0
Recommended Storage : 0° to 5°C
- Carbenicillin disodium salt
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.