Learn More
Corning™ Carbenicillin, Disodium Salt, Powder
Store: 2° to 8°C
Brand: Corning™ 46-100-RG
Code : NEW
Additional Details : CAS Number : 4800-94-6 Weight : 0.08100kg
Specifications
Powder | |
Antibiotic | |
Plastic Bottle | |
1 x 5 g |
Store: 2° to 8°C
Brand: Corning™ 46-100-RG
Code : NEW
Additional Details : CAS Number : 4800-94-6 Weight : 0.08100kg
Powder | |
Antibiotic | |
Plastic Bottle | |
1 x 5 g |