Learn More
Carbon black, acetylene, 100% compressed, Thermo Scientific Chemicals
£49.10 - £144.00
Chemical Identifiers
CAS | 1333-86-4 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.01 |
MDL Number | MFCD00133992 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15496985
|
Thermo Scientific Alfa Aesar
045527.30 |
250 g |
£49.10
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15406995
|
Thermo Scientific Alfa Aesar
045527.95 |
2 x 500 g |
£144.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in optical computing, optical memories, optical signal processing and controlling applications respects, C60 and its derivatives can be widely used in photovoltaic applications, magnetic resonance imaging, anti-HIV drugs, anti-cancer drugs, chemotherapy drugs, cosmetic additives, research and other areas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
1333-86-4 | |
12.01 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N |
C | |
MFCD00133992 |
Specifications
1333-86-4 | |
1.8 to 2.1g/cm3 at 20°C | |
C | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
Powder |
Black | |
>99.9% | |
MFCD00133992 | |
12.01 | |
Carbon black |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H351
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III
missing translation for 'einecsNumber' : 215-609-9
missing translation for 'rtecsNumber' : FF5800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only