Learn More
Azpack™ Carbon Chrome Steel Spheres
Demonstrates Archimedes' Principle and other properties of physics and math. Azpack™ Carbon Chrome Steel Spheres are used mainly for educational purposes.
Brand:  Azpack™ 12403090
 View more versions of this product
Product Code. 12403090
  • £31.00 / Pack of 25
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Manufactured from carbon chrome steel.
  • Available in a variety of sizes
  • Smooth, polished chrome surface
  • Round-shaped balls
Specifications
Specifications
Metal Ball
Carbon Chrome Steel
Dia: 12mm
12 mm