missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Dioxide Standard (CO2) 3 g/L, Reagecon
One of a series of standards used to calibrate and control the Corning 965D Carbon Dioxide Analyzer.
Brand: Reagecon SC023C
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- For calibrating carbon dioxide analyzer.
- Useful for determining carbon dioxide content in carbonated beverages.
- Uses raw materials of very high purity and specification.
- Tested titremetrically using Reagecon's in-house method TPATB against hydrochloric acid, analytical volumetric standard solution.
- Standard is directly traceable to NIST Standard Reference Material 723e - tris(hydroxymethyl)aminomethane.
- A detailed Certificate of Analysis is issued with each batch manufactured.
Specifications
Carbon Dioxide Analysis | |
100 x 2 mL, 3 g/L |