Learn More
Carbon Dioxide Standard (CO2) Release Agent, Reagecon
Used as an acidic CO2 release agent during the measurement of CO2 using the Corning 965D Carbon Dioxide Analyzer
Brand: Reagecon RR021
Code : 03
Additional Details : CAS Number : 598-82-3 Weight : 1.44000kg
Description
- Use of this instrument is one of a number of approved techniques available for the determination of carbon dioxide, in beer and other carbonated beverages.
- This instrument was subsequently marketed by Mettler Toledo as the model M965D CO2 Analyzer.
- Produced using raw materials of a very high purity and specification.
- Tested using Reagecon Test Method TPATA with results expressed in molar acidity, specification 2.0 – 2.2 M acidity.
- A Certificate of Conformity is issued with each batch.
Specifications
Carbon Dioxide Analysis | |
6 x 200 mL |
Safety and Handling
- CO2 RELEASE REAGENT
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
- MIXTURE LIST-Contains : Lactic acid