missing translation for 'onlineSavingsMsg'
Learn More

Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals

Product Code. 13418598
1 L, Glass Bottle
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-15-
Molecular Formula CS2
Molecular Weight (g/mol) 76.13

Specifications

Melting Point -111.0°C
Density 1.2620g/mL
Boiling Point 46.0°C
Quantity 1 L
Flash Point −30°C
Packaging Glass Bottle
Refractive Index 1.627
Assay Percent Range 99.85% min. (GC)
Specific Gravity 1.262
Linear Formula CS2
Viscosity 0.363 mPa.s (20°C)
Merck Index 151811
Solubility Information Solubility in water: 2.9g/L (20°C). Other solubilities: miscible with anhydrous methanol,ethanol,ether,,benzene,chloroform,carbon tetrachloride and oils
Formula Weight 76.13
Percent Purity 99.9%
Grade HPLC
Chemical Name or Material Carbon disulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465292 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog Number445660010,445661000,445660025
Lot NumberA0465293 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number445660010,445661000,445660025
Lot NumberA0462822 Certificate TypeCertificate of Analysis Date14/05/2024 Catalog Number445660010,445661000,445660025
Lot NumberA0443815 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog Number445660010,445661000,445660025
Lot NumberA0458176 Certificate TypeCertificate of Analysis Date30/11/2023 Catalog Number445660010,445661000,445660025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Carbon disulfide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P362-Take off contaminated clothing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Carbon disulfide, 99.9%, for HPLC, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.