missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon disulfide, ACS Reagent, ≥99.9%, Honeywell Riedel-de Haën™
ACS Reagent,≥99.9%
£105.73 - £197.46
Chemical Identifiers
CAS | 75-15-0 |
---|---|
Molecular Formula | CS2 |
Molecular Weight (g/mol) | 76.131 |
MDL Number | MFCD00011321 |
InChI Key | QGJOPFRUJISHPQ-UHFFFAOYSA-N |
Synonym | carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide |
PubChem CID | 6348 |
ChEBI | CHEBI:23012 |
IUPAC Name | methanedithione |
SMILES | C(=S)=S |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15656700
|
Honeywell-Riedel-de Haen
676918-1L |
1 L | Glass Bottle |
£105.73
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15678450
|
Honeywell-Riedel-de Haen
676918-4L |
4 L | Glass Bottle | N/A | |||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
75-15-0 | |
76.131 | |
QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
6348 | |
methanedithione |
CS2 | |
MFCD00011321 | |
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
CHEBI:23012 | |
C(=S)=S |
Specifications
-76°C | |
Colorless | |
46°C (1013 hPa) | |
212°C | |
Liquid | |
MFCD00011321 | |
1098293 | |
2.9g/L in water at 20°C | |
C(=S)=S | |
76.131 | |
CHEBI:23012 | |
≥99.9% | |
Carbon disulfide |
75-15-0 | |
1.266g/mL at 25°C | |
0.5 | |
Unpleasant | |
CS2 | |
UN1131 | |
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
methanedithione | |
6348 | |
76.14g/mol | |
ACS Reagent |
Safety and Handling
P201-P210-P261-P280-P304 + P340 + P312-P308 + P313
missing translation for 'dotInformation' : class 3 - PG 1 - Carbon disulfide
missing translation for 'rtecsNumber' : 200-843-6
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Carbon disulfide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : carbon disulphide