missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Disulfide, ACS Reagent, ≥99.9%, Honeywell Riedel-de Haën™
ACS reagent,≥99.9%
£74.02 - £133.78
Chemical Identifiers
CAS | 75-15-0 |
---|---|
Molecular Formula | CS2 |
Molecular Weight (g/mol) | 76.131 |
MDL Number | MFCD00011321 |
InChI Key | QGJOPFRUJISHPQ-UHFFFAOYSA-N |
Synonym | carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide |
PubChem CID | 6348 |
ChEBI | CHEBI:23012 |
IUPAC Name | methanedithione |
SMILES | C(=S)=S |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15631650
|
Honeywell-Riedel-de Haen
180173-500ML |
500 mL | Glass Bottle |
£74.02
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15621650
|
Honeywell-Riedel-de Haen
180173-1L |
1 L | Glass Bottle |
£133.78
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
75-15-0 | |
76.131 | |
QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
6348 | |
methanedithione |
CS2 | |
MFCD00011321 | |
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
CHEBI:23012 | |
C(=S)=S |
Specifications
-76°C | |
1.266g/mL at 25°C | |
0.5 | |
212°F | |
n20/D 1.627(lit.) | |
MFCD00011321 | |
1098293 | |
QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
methanedithione | |
6348 | |
76.14g/mol | |
ACS Reagent |
75-15-0 | |
46°C | |
−30°C (−22°F) | |
Liquid | |
CS2 | |
UN1131 | |
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
C(=S)=S | |
76.131 | |
CHEBI:23012 | |
≥99.9% | |
Carbon Disulfide |
Safety and Handling
P210-P260-P280-P305 + P351 + P338-P370 + P378-P403 + P235
H225-H315-H319-H332-H361fd-H372
missing translation for 'dotInformation' : Class 3 - PG 1 - Carbon disulfide
missing translation for 'einecsNumber' : 200-843-6
missing translation for 'rtecsNumber' : FF6650000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Carbon disulfide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : carbon disulphide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title