missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Carbon Filters for Protector™™ Workstations

Absorb harmful chemical fumes and vapors

Brand:  Labconco™ 3924201

Additional Details : CAS Number : 7681-11-0 Weight : 0.80000kg

 View more versions of this product

Product Code. 11804401

  • £565.00 / Each
Estimated Shipment: 18-11-2024
to see stock.

Description

Description

Compatible with Protector Work Stations with Built-In Blowers (2004 or newer).

Order Info

18W x 36D x 1 in.H (45.7 x 914 x 2.5cm).

Specifications

Specifications

Carbon
1 in.
6.4 Kg
36 in.
2.5 cm
91.4 cm
45.7 cm
Adsorb Formalin, Formaldehyde
Protector Work Station
14 lb.
30 to 40 in.
18 in.
30 to 40 x 18 x 1 in. (76.2 to 101.6 x 45.7 x 2.5 cm)
76.2 to 101.6 cm
Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers