Learn More
Swann-Morton™ Carbon Steel Individual Foil Packs, Sterile Scalpel Blades, No. 9
Distinctive, broad, hatchet-shaped blade.
Brand:  Swann-Morton™ 0217
Additional Details : Weight : 0.01000kg
Product Code. 15957682
  • £22.00 / Pack of 100
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
  • Broad, hatchet-shaped.
  • Used within podiatry.
  • Fitment No. 3.
  • Fits handles 3, 3L, 3 graduated, 5B, 7, 9, B3, and B3L.
Specifications
Specifications
Carbon Steel
Sterile
100/Box
Individually Packaged