Learn More
Swann-Morton™ Carbon Steel Individual Peel Pack, Non-Sterile Scalpel Blades, No. 11
Product Code. 15803252
Click to view available options
Quantity:
100/Box
Unit Size:
Pack of 100
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15803252
Brand: Swann-Morton™ 3003
Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Elongated triangular blade sharpened along the hypotenuse edge with a strong pointed tip making it ideal for stab incisions.
  • Elongated triangular blade.
  • Sharpened along the hypotenuse edge.
  • Strong, pointed tip, making it ideal for stab incisions needed when lancing an abscess or inserting a chest drain.
  • It is held like a pencil and often upside down by the surgeon to prevent it inadvertently being inserted too deeply.
  • To avoid this scenario, the scalpel should not be held towards the back end whilst the heel of the hand should be placed on the skin thus stabilizing the hand to help prevent an unwanted deep incision.
  • The No.11 is also used for other small incisions such as when inserting a laparoscopic port, vein and artery central lines, opening the aorta and removing calcifications in the aortic or mitral valves, anatomical dissections, and surgical treatment of disorders of the foot, ankle, and lower extremities in podiatry.
  • Fits handles 3, 3L, 3 Graduated, 5B, 7, 9 and B3.
Specifications
Blade Material Carbon Steel
Sterility Non-sterile
Quantity 100/Box
Packaging Type Individually Packaged