Learn More
Azpack™ Carbon Steel Razor Blades
Packaged in drawer box merchandiser. Azpack™ Carbon Steel Razor Blades are individually wrapped in paper cardstock for safety.
Brand:  Azpack™ 11904325
Product Code. 11904325
  • £128.00 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Packaged in drawer box merchandiser. Azpack™ Carbon Steel Razor Blades are individually wrapped in paper cardstock for safety.
  • Carbon steel
  • Regular duty
  • Single-edged blade
  • Contains aluminum spine
  • Individually wrapped
Specifications
Specifications
Carbon Steel
38.1 x 19 mm
10/Pk.
Razor Blade
0.22 mm