Learn More
Swann-Morton™ Carbon Steel Sterile Scalpel Blades
Use for convenience in user environments: on the ward, in emergency or trauma units or even out in the field with paramedics. Swann Morton™ Carbon Steel Surgical Scalpel Blades come in single peel packs.
Brand: Swann-Morton™ 0211
This item is not returnable.
View return policy
Description
- Carbon steel to BS EN 27740
- Sterile
Specifications
Carbon Steel | |
Used for Making Long Incisions in General Surgery and also in Autopsy Procedures | |
The # 24 fits Handles # 4, # 4L, # 4 Graduated and # 6B | |
CE compliant 93/42/EC |
Scalpel Blade | |
# 24 | |
Sterile | |
10/Pk. |