missing translation for 'onlineSavingsMsg'
Learn More

Carbon yarn, woven from 0.076mm (0.003in) dia fibers, Thermo Scientific Chemicals

£27.54 - £96.98

Chemical Identifiers

MDL Number MFCD00133992
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364677
View Documents
Thermo Scientific Alfa Aesar
039721.G5
5 m
£27.54
5m
Please to purchase this item. Need a web account? Register with us today!
11374677
View Documents
Thermo Scientific Alfa Aesar
039721.G9
25 m
£96.98
25m
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00133992

Specifications

MFCD00133992
Carbon yarn, Woven from 0.076mm (0.003 in.) dia. fibers
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-153-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only