missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carbonate, 0.5M buffer soln., pH 9.6

Brand:  Thermo Scientific Chemicals J62610.AP

104.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15424949

  • £104.52 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Sodium bicarbonate buffer system is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it. It is used to neutralize the pH of the chyme leaving the stomach and entering the small intestine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bicarbonate buffer system is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it. It is used to neutralize the pH of the chyme leaving the stomach and entering the small intestine.

Solubility
It is soluble in water.

Notes
Store at 4°C in tightly closed container. Avoid inhalation. Wash thoroughly after handling. Avoid prolonged or repeated exposure.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.5 M buffer soln.
9.6
500 mL
Carbonate buffer soln.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW16K536 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog NumberJ62610.AP,J62610.AK
Lot NumberU01K556 Certificate TypeCertificate of Analysis Date05/08/2024 Catalog NumberJ62610.AP,J62610.AK
Lot NumberS01K528 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog NumberJ62610.AP,J62610.AK
Lot NumberP24K509 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ62610.AP,J62610.AK
Lot NumberQ04K536 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog NumberJ62610.AP,J62610.AK
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Carbonate, 0.5M buffer soln., pH 9.6 > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only