missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonate solution, 0.18M Carb./0.17M Bicarb. for Ion Chromatography, 100mg/L, H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£68.70 - £125.00
Specifications
Concentration | 100mg/L |
---|---|
Type | Single-component Standard Solution |
Solution Contains | 0.18M Carbonate/0.17M Bicarbonate |
Solvent or Matrix | H2O |
Physical Form | Liquid |
Chemical Name or Material | Carbonate solution |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12902245
|
Fisher Chemical
A 105 100 |
100 mL | Bottle |
£68.70
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12912245
|
Fisher Chemical
A 105 500 |
500 mL | Bottle |
£125.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
100mg/L | |
0.18M Carbonate/0.17M Bicarbonate | |
Liquid |
Single-component Standard Solution | |
H2O | |
Carbonate solution |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)