missing translation for 'onlineSavingsMsg'
Learn More

Carbonate solution, 0.5M Sodium Carbonate, for Ion Chromatography, H2O matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

£75.75 - £128.00

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12962235
View Documents
Fisher Chemical
A 103 100
100 mL Bottle
£75.75
100mL
Please to purchase this item. Need a web account? Register with us today!
12972235
View Documents
Fisher Chemical
A 103 500
500 mL Bottle
£128.00
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

497-19-8
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
CHEBI:29377
CNa2O3
MFCD00003494
10340
[Na+].[Na+].[O-]C([O-])=O

Specifications

0.5 M
Single-component Standard Solution
MFCD00003494
[Na+].[Na+].[O-]C([O-])=O
105.99
CHEBI:29377
H2O
Carbonate
497-19-8
CNa2O3
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium;carbonate
10340
0.5M Sodium Carbonate
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months. (12 Months after opening)

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.