missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Carbonic Anhydrase (Bovine)
Used for Enzyme
Brand: Abnova™ P5208.50mg
Additional Details : Weight : 0.00010kg
Description
#REF!Specifications
P00921 | |
510801 | |
Carbonic Anhydrase (Bovine) | |
50 mg | |
RUO | |
CA1 | |
≥3,000 units per mg dry weight. One Unit is the time required (in seconds) for a saturated CO2 solution to lower the pH of 0.02M Tris-HCl buffer from 8.3 to 6.3, at 0-4°C is determined. | |
None | |
Lyophilized |
Enzyme Activity | |
29kDa | |
Native protein purified from Bovine Erythrocytes | |
Store at 4°C. | |
CA1 | |
Carbonic Anhydrase Assay; CO | |
Bovine | |
Native protein purified from Bovine Erythrocytes |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 50MG Carbonic Anhydrase
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic