Learn More
Carbonic Anhydrase I Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547264
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, less than 5% cross‑reactivity with recombinant human (rh) CA2, rhCA3, rhCA4, rhCA8, rhCA9, rhCA10, and rhCA12 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Carbonic Anhydrase I Polyclonal antibody specifically detects Carbonic Anhydrase I in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
Carbonic Anhydrase I | |
Polyclonal | |
Unconjugated | |
CA1 | |
AW555628; CA I; CA1; ca1 {ECO:0000250; CAB; CA-I; CA-I {ECO:0000250; Car1; Car-1; Carbonate dehydratase I; carbonate dehydratase I {ECO:0000250; carbonic anhydrase 1; carbonic anhydrase 1 {ECO:0000250; Carbonic anhydrase B; carbonic anhydrase I; carbonic anhydrase I {ECO:0000250; epididymis secretory protein Li 11; HEL-S-11; UniProtKB:P00915} | |
Goat | |
Antigen affinity chromatography | |
RUO | |
759 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P00915 | |
CA1 | |
E. coli-derived recombinant human Carbonic Anhydrase I/CA1 Ala2-Phe261 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |