missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase III/CA3 Antibody Pair, Novus Biologicals™
Antibody Pair
Brand: Novus Biologicals H00000761-AP11
Additional Details : Weight : 0.00970kg
Description
Carbonic Anhydrase III/CA3 antibody specifically detects Carbonic Anhydrase III/CA3 in Human samples. It is validated for ELISASpecifications
Carbonic Anhydrase III/CA3 | |
Unconjugated | |
1x PBS, pH 7.4 | |
1 Set | |
Lipid and Metabolism | |
761 | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
Sandwich ELISA | |
Sandwich ELISA 3 to 100 ng/mL | |
CAIII, CA-III, Car3, Carbonate dehydratase III, carbonic anhydrase 3, Carbonic anhydrase III, carbonic anhydrase III, muscle specific, EC 4.2.1.1 | |
RUO | |
Primary | |
Human | |
Purified |