missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase III Monoclonal Antibody (5G8B6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA542991
Additional Details : Weight : 0.01000kg
Description
Specificity: Human Carbonic Anhydrase III/CA3
Carbonic anhydrase III (CAIII) is a member of a multigene family (at least six separate genes are known) that encodes carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal. The gene spans 10. 3 kb and contains seven exons and six introns.Specifications
Carbonic Anhydrase III | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P07451 | |
Ca3 | |
Recombinant Human Carbonic Anhydrase III/CA3 Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
5G8B6 | |
Unconjugated | |
Ca3 | |
BB219044; CA III; Ca3; CAH3; CAIII; CA-III; CAIII (Muscle); Car3; Car-3; Carbonate dehydratase III; carbonic anhydrase 3; Carbonic anhydrase III; carbonic anhydrase III, muscle specific; carbonic anhydrase-like protein; EC 4.2.1.1; epididymis secretory sperm binding protein Li 167mP; HEL-S-167mP | |
Mouse | |
Protein A | |
RUO | |
761 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |