missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase III Monoclonal Antibody (5G8B6), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA543509

 View more versions of this product

Product Code. 17779853

  • £199.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specificity: Human Carbonic Anhydrase III/CA3

Carbonic anhydrase III (CAIII) is a member of a multigene family (at least six separate genes are known) that encodes carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal. The gene spans 10. 3 kb and contains seven exons and six introns.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carbonic Anhydrase III
Monoclonal
1 mg/mL
PBS with no preservative
P07451
Ca3
Recombinant Human Carbonic Anhydrase III/CA3 Protein.
50 μL
Primary
Human
Antibody
IgG1
ELISA
5G8B6
Unconjugated
Ca3
BB219044; CA III; Ca3; CAH3; CAIII; CA-III; CAIII (Muscle); Car3; Car-3; Carbonate dehydratase III; carbonic anhydrase 3; Carbonic anhydrase III; carbonic anhydrase III, muscle specific; carbonic anhydrase-like protein; EC 4.2.1.1; epididymis secretory sperm binding protein Li 167mP; HEL-S-167mP
Mouse
Protein A
RUO
761
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers