missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase III Monoclonal Antibody (C2), Invitrogen™
Mouse Monoclonal Antibody
£146.00 - £329.00
Specifications
Antigen | Carbonic Anhydrase III |
---|---|
Clone | C2 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17918782
|
Invitrogen
MA550993 |
200 μL |
£329.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17959582
|
Invitrogen
MA550994 |
20 μL |
£146.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbonic anhydrase III (CAIII) is a member of a multigene family (at least six separate genes are known) that encodes carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal. The gene spans 10. 3 kb and contains seven exons and six introns.Specifications
Carbonic Anhydrase III | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4 | |
BB219044, CA III, Ca3, CAH3, CAIII, CA-III, CAIII (Muscle), Car3, Car-3, Carbonate dehydratase III, carbonic anhydrase 3, Carbonic anhydrase III, carbonic anhydrase III, muscle specific, carbonic anhydrase-like protein, EC 4.2.1.1, epididymis secretory sperm binding protein Li 167mP, HEL-S-167mP | |
Ca3, Car3 | |
IgG2b κ | |
Protein A/G | |
Ca3 |
C2 | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Porcine, Rat | |
P07451, P14141, P16015, Q5S1S4 | |
12350, 494016, 54232, 761 | |
Recombinant Human CA3 protein, Ala2-Lys260 (Accession #P07451) | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |