Learn More
Carbonic Anhydrase IV Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547312
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant human (rh) CA4 is observed and less than 2% cross-reactivity with rhCA3, rhCA10, rhCA13, recombinant mouse (rm) CA9, and rmCA12 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Carbonic Anhydrase IV Polyclonal antibody specifically detects Carbonic Anhydrase IV in Mouse samples. It is validated for Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
Carbonic Anhydrase IV | |
Polyclonal | |
Unconjugated | |
Car4 | |
AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
12351 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q64444 | |
Car4 | |
Mouse myeloma cell line NS0-derived recombinant mouse Carbonic Anhydrase IV/CA4 Glu18-Ser277 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |