missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IV Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
£146.00 - £337.00
Specifications
Antigen | Carbonic Anhydrase IV |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17910023
|
Invitrogen
PA5145356 |
20 μL |
£146.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17939462
|
Invitrogen
PA5145355 |
200 μL |
£337.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known, however, it may have a role in inherited renal abnormalities of bicarbonate transport.Specifications
Carbonic Anhydrase IV | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Porcine, Rat | |
P22748, P48284, Q64444 | |
100511956, 12351, 29242, 762 | |
Recombinant Rat CA4 protein, Ser20-Ser220 (Accession #P48284) | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4 | |
AW456718, CA IV, CA4, CAIV, CA-IV, Car4, Carbonate dehydratase IV, carbonic anhydrase 4, Carbonic anhydrase IV, carbonic dehydratase IV, RP17 | |
CA4, Car4 | |
IgG | |
Affinity chromatography | |
CA4 |