missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IV Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542002
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: DGEHFAMEMH IVHEKEKGTS RNVKEAQDPE DEIAVLAFLV EAGTQVNEGF Sequence homology: Human: 100%
Carbonic Anhydrase IV Polyclonal antibody specifically detects Carbonic Anhydrase IV in Human samples. It is validated for Western Blot.Specifications
Carbonic Anhydrase IV | |
Polyclonal | |
Unconjugated | |
CA4 | |
AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17 | |
Rabbit | |
Protein A | |
RUO | |
762 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P22748 | |
CA4 | |
Synthetic peptide directed towards the middle region of human CA4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |