missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase VA Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA5145353

 View more versions of this product

Product Code. 17989522

  • £337.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric juice. They show extensive diversity in distribution and in their subcellular localization. The human CA5A gene encodes for the CA V protein which is expressed in the mitochondrial matrix in liver tissues, but can also be detected in many other tissues. In ureagenesis, the CA V protein may be involved in providing bicarbonate to carbamoyl phosphate synthetase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carbonic Anhydrase VA
Polyclonal
Unconjugated
Car5a
CA Y, CA5, CA5A, CA5AD, Car5, Car5a, Carbonate dehydratase VA, carbonic anhydrase 5 mitochondrial, carbonic anhydrase 5, mitochondrial, carbonic anhydrase 5A, carbonic anhydrase 5a, mitochondrial, carbonic anhydrase 5b, mitochondrial, carbonic anhydrase V, mitochondrial, carbonic anhydrase VA, carbonic anhydrase VA, mitochondrial, carbonic dehydratase, CAV, CAVA, CA-VA, GS1-21A4.1
Rabbit
Protein A
RUO
12352, 54233
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4
P23589, P43165
CA5A, Car5a
Recombinant Rat CA5A protein, Tyr12-Leu219 (Accession #P43165)
200 μL
Primary
Mouse, Rat
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers