missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase VI Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

£146.00 - £337.00

Specifications

Antigen Carbonic Anhydrase VI
Concentration 0.5 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17920543
View Documents
Invitrogen
PA5145390
20 μL
£146.00
20µL
Please to purchase this item. Need a web account? Register with us today!
17950653
View Documents
Invitrogen
PA5145389
200 μL
£337.00
200µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is one of several isozymes of carbonic anhydrase. This protein is found only in salivary glands and saliva and protein may play a role in the reversible hydratation of carbon dioxide though its function in saliva is unknown.
Specifications

Specifications

Carbonic Anhydrase VI
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Rat
CA6, CA-6, Car6, Carbonate dehydratase VI, carbonic anhydrase 6, Carbonic anhydrase VI, carbonic anhydrase VI nirs variant 2, Carbonic anhydrase6, CA-VI, DOC1, GUSTIN, salivary carbonic anhydrase, Secreted carbonic anhydrase
Car6
IgG
Protein A
Car6
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4
298657
Recombinant Rat CA6 protein, Thr147-Ala275 (Accession #F1LQ08)
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications