Learn More
Carbonic Anhydrase VIII Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528555
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa. Predicted reactivity: Zebrafish (83%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene was initially named CA-related protein because of sequence similarity to other known carbonic anhydrase genes. However, the gene product lacks carbonic anhydrase activity (i.e., the reversible hydration of carbon dioxide). The gene product continues to carry a carbonic anhydrase designation based on clear sequence identity to other members of the carbonic anhydrase gene family. The absence of CA8 gene transcription in the cerebellum of the lurcher mutant in mice with a neurologic defect suggests an important role for this acatalytic form.Specifications
Carbonic Anhydrase VIII | |
Polyclonal | |
Unconjugated | |
Ca8 | |
AW546993; Ca8; Cals; Cals1; CAMRQ3; Car8; carbonate dehydratase; carbonic anhydrase 8; carbonic anhydrase VIII; carbonic anhydrase-like sequence; carbonic anhydrase-like sequence 1; carbonic anhydrase-related protein; CA-related protein; CARP; CA-RP; CARP VIII; CA-RP VIII; CA-VIII; hypothetical protein LOC550233; LOW QUALITY PROTEIN: carbonic anhydrase-related protein; MGC120502; MGC99509; wdl; zgc:110118 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
767 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P35219 | |
Ca8 | |
Recombinant fragment corresponding to a region within amino acids 5 and 290 of Human CA8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |